siRNA Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122738.1 | NP_001116210.1 | 14 | 2796 | ||
| NM_177645.4 | NP_808313.3 | 13 | 2769 | |||
| GenBank mRNA | AK082897.1 | - | 13 | 1616 | ||
| AK220534.1 | - | 13 | 2720 | |||
| BC141036.1 | - | 14 | 2679 | 9055648 | ||

Kansl1l
Genomic Map