siRNA Details
| Gene Symbol: Mrps18c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026826.1 | NP_081102.1 | 3 | 229 | ||
| GenBank mRNA | AK004139.1 | - | 3 | 242 | ||
| AK088290.1 | - | 3 | 242 | |||
| AK145935.1 | - | 3 | 246 | |||
| BC028442.1 | - | 3 | 229 | 1196057 | ||

Mrps18c
Genomic Map