siRNA Details
| Gene Symbol: Tyw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037742.2 | NP_001032831.2 | 2 | 413 | ||
| GenBank mRNA | AK139666.1 | - | 2 | 277 | ||
| BC023868.1 | - | 2 | 424 | |||
| BC058590.1 | - | 2 | 189 | |||
| BC099495.1 | - | 2 | 174 | |||
| BC132289.1 | - | 2 | 262 | 40130566 | ||
| BC144765.1 | - | 2 | 262 | |||
| BC144766.1 | - | 2 | 262 | |||
| BU697704.1 | - | 2 | 380 | |||
| CF951173.1 | - | 2 | 183 | |||

Tyw5
Genomic Map