siRNA Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037742.2 | NP_001032831.2 | 4 | 532 | ||
GenBank mRNA | AK139666.1 | - | 4 | 377 | ||
BC023868.1 | - | 4 | 524 | |||
BC058590.1 | - | 4 | 289 | |||
BC099495.1 | - | 4 | 274 | |||
BC132289.1 | - | 4 | 381 | 40130566 | ||
BC144765.1 | - | 4 | 362 | |||
BC144766.1 | - | 4 | 362 | |||
CF951173.1 | - | 4 | 283 |
