siRNA Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026827.1 | NP_081103.1 | 2, 3 | 265 | ||
| NM_028389.1 | NP_082665.1 | 2, 3 | 262 | |||
| GenBank mRNA | AK004043.1 | - | 1, 2 | 225 | ||
| AK152526.1 | - | 2, 3 | 227 | |||
| BC023442.1 | - | 2, 3 | 240 | 5026775 | ||
| BC046763.1 | - | 1, 2 | 407 | 5693779 | ||
| BY128578.1 | - | 2, 3 | 265 | |||

Tmem219
Genomic Map