siRNA Details
| Gene Symbol: Zfp740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289690.1 | NP_001276619.1 | 5, 6 | 1025 | ||
| NM_001289691.1 | NP_001276620.1 | 7, 8 | 1224 | |||
| NM_001289694.1 | NP_001276623.1 | 6, 7 | 933 | |||
| NM_001289695.1 | NP_001276624.1 | 6, 7 | 899 | |||
| GenBank mRNA | AK144707.1 | - | 7, 8 | 1221 | ||
| AK170207.1 | - | 6, 7 | 921 | |||
| BC030410.1 | - | 2, 3 | 730 | 5367159 | ||
| BC066011.1 | - | 6, 7 | 793 | 6418561 | ||
| CA319591.1 | - | 5, 6 | 563 | |||

Zfp740
Genomic Map