siRNA Details
| Gene Symbol: 1110038B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002372.1 | - | 3 | 180 | ||
| AK003073.1 | - | 3 | 178 | |||
| AK004150.1 | - | 3 | 178 | |||
| BC058123.1 | - | 3 | 164 | |||
| BE133139.1 | - | 2 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110038B12Rik| Gene Symbol: 1110038B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002372.1 | - | 3 | 180 | ||
| AK003073.1 | - | 3 | 178 | |||
| AK004150.1 | - | 3 | 178 | |||
| BC058123.1 | - | 3 | 164 | |||
| BE133139.1 | - | 2 | 267 | |||