siRNA Details
| Gene Symbol: Cdhr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024478.1 | NP_001019649.1 | 8 | 1016 | ||
| GenBank mRNA | AK004207.1 | - | 2 | 199 | ||
| AK047666.1 | - | 8 | 1017 | |||
| AK049806.1 | - | 8 | 1018 | |||
| BC138911.1 | - | 8 | 944 | 8861933 | ||

Cdhr3
Genomic Map