siRNA Details
| Gene Symbol: Phtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172992.3 | NP_766580.3 | 17 | 2407 | ||
| GenBank mRNA | AK037021.1 | - | 17 | 2294 | ||
| AK162226.1 | - | 6 | 852 | |||
| AK163376.1 | - | 17 | 2334 | |||
| BC052841.1 | - | 17 | 2320 | 30050876 | ||

Phtf2
Genomic Map