siRNA Details
| Gene Symbol: Flnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081185.1 | NP_001074654.1 | 38 | 6633 | ||
| GenBank mRNA | AK135523.1 | - | 3 | 473 | ||
| BC039885.1 | - | 3 | 501 | |||
| BC052186.1 | - | 11 | 1582 | |||
| BC060276.1 | - | 17 | 2694 | |||
| BC151097.1 | - | 38 | 6438 | 9088010 | ||
| BC158128.1 | - | 37 | 6339 | 9007396 | ||
| BC172159.1 | - | 37 | 6339 | |||

Flnc
Genomic Map