siRNA Details
| Gene Symbol: Prr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026841.1 | NP_081117.1 | 2 | 358 | ||
| GenBank mRNA | AK004321.1 | - | 1 | 213 | ||
| BC060289.1 | - | 2 | 358 | |||
| BC094615.1 | - | 2 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr32| Gene Symbol: Prr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026841.1 | NP_081117.1 | 2 | 358 | ||
| GenBank mRNA | AK004321.1 | - | 1 | 213 | ||
| BC060289.1 | - | 2 | 358 | |||
| BC094615.1 | - | 2 | 326 | |||