siRNA Details
| Gene Symbol: Dock5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177780.3 | NP_808448.2 | 8 | 915 | ||
| GenBank mRNA | AK053909.1 | - | 8 | 786 | ||
| AK147975.1 | - | 8 | 776 | |||
| BC158046.1 | - | 8 | 775 | 9007351 | ||
| BC158135.1 | - | 8 | 775 | 9088200 | ||
| BC158138.1 | - | 8 | 775 | 9088202 | ||

Dock5
Genomic Map