siRNA Details
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133700.2 | NP_598461.1 | 8 | 1105 | ||
GenBank mRNA | AK054369.1 | - | 2 | 1522 | ||
BC050781.1 | - | 8 | 1105 | 6591397 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btbd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133700.2 | NP_598461.1 | 8 | 1105 | ||
GenBank mRNA | AK054369.1 | - | 2 | 1522 | ||
BC050781.1 | - | 8 | 1105 | 6591397 |