siRNA Details
| Gene Symbol: Ddi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017966.2 | NP_001017966.1 | 2, 3 | 484 | ||
| GenBank mRNA | AK129015.1 | - | 464 | |||
| BC021415.1 | - | 2, 3 | 465 | |||
| BY731719.1 | - | 2, 3 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddi2| Gene Symbol: Ddi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017966.2 | NP_001017966.1 | 2, 3 | 484 | ||
| GenBank mRNA | AK129015.1 | - | 464 | |||
| BC021415.1 | - | 2, 3 | 465 | |||
| BY731719.1 | - | 2, 3 | 484 | |||