siRNA Details
| Gene Symbol: Pih1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278207.1 | NP_001265136.1 | 3 | 246 | ||
| NM_001285904.1 | NP_001272833.1 | 3 | 238 | |||
| NM_029406.4 | NP_083682.1 | 3 | 366 | |||
| GenBank mRNA | AK004341.1 | - | 3 | 247 | ||
| AK016797.1 | - | 3 | 272 | |||
| AK089003.1 | - | 3 | 315 | |||
| AK146039.1 | - | 3 | 304 | |||
| AK152695.1 | - | 3 | 235 | |||
| AK177389.1 | - | 2 | 111 | |||
| BC068254.1 | - | 3 | 255 | 30013096 | ||

Pih1d1
Genomic Map