siRNA Details
| Gene Symbol: Asb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026853.1 | NP_081129.1 | 2 | 169 | ||
| GenBank mRNA | AF398972.1 | - | 2 | 283 | ||
| AK002412.1 | - | 2 | 256 | |||
| AK004396.1 | - | 2 | 169 | |||
| AK005404.1 | - | 2 | 283 | |||
| AK078851.1 | - | 3 | 481 | |||
| AK146664.1 | - | 2 | 283 | |||
| BC064047.1 | - | 2 | 165 | 864428 | ||

Asb11
Genomic Map