siRNA Details
Gene Symbol: Dtwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026854.3 | NP_081130.1 | 4, 5 | 656 | ||
GenBank mRNA | AK004450.1 | - | 4, 5 | 631 | ||
AK144689.1 | - | 4, 5 | 631 | |||
BC075693.1 | - | 6, 7 | 750 | |||
BC139347.1 | - | 4, 5 | 622 | 8862369 | ||
BC139348.1 | - | 4, 5 | 622 | 8862370 |
