siRNA Details
| Gene Symbol: Klhdc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033039.2 | NP_001028211.1 | 4 | 1106 | ||
| GenBank mRNA | AK004455.1 | - | 1 | 489 | ||
| AK076812.1 | - | 4 | 829 | |||
| AK140290.1 | - | 4 | 1107 | |||
| BC141196.1 | - | 4 | 1019 | 9055808 | ||

Klhdc9
Genomic Map