siRNA Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133701.2 | NP_598462.1 | 2 | 254 | ||
| GenBank mRNA | AK011639.1 | - | 2 | 196 | ||
| AK081998.1 | - | 2 | 189 | |||
| AK145461.1 | - | 2 | 257 | |||
| AK166890.1 | - | 2 | 206 | |||
| BC014869.1 | - | 2 | 181 | 3602158 | ||
| BC023691.1 | - | 2 | 183 | 5344332 | ||
| BC025030.1 | - | 2 | 200 | 4947416 | ||
| BY735790.1 | - | 2 | 254 | |||

Prpf6
Genomic Map