siRNA Details
| Gene Symbol: Rasl11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026864.1 | NP_081140.1 | 4 | 503 | ||
| GenBank mRNA | AK004371.1 | - | 4 | 457 | ||
| BC118009.1 | - | 1618 | 40102480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasl11a| Gene Symbol: Rasl11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026864.1 | NP_081140.1 | 4 | 503 | ||
| GenBank mRNA | AK004371.1 | - | 4 | 457 | ||
| BC118009.1 | - | 1618 | 40102480 | |||