siRNA Details
| Gene Symbol: Tatdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163421.1 | NP_001156893.1 | 4, 5 | 335 | ||
| NM_001163422.1 | NP_001156894.1 | 4, 5 | 335 | |||
| NM_026895.1 | NP_081171.1 | 4, 5 | 335 | |||
| GenBank mRNA | AK005193.1 | - | 4, 5 | 306 | ||
| AK156071.1 | - | 4, 5 | 286 | |||
| BC061248.1 | - | 4, 5 | 307 | |||

Tatdn3
Genomic Map