siRNA Details
| Gene Symbol: Anapc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181394.3 | NP_852059.1 | 2 | 108 | ||
| GenBank mRNA | AK007332.1 | - | 1 | 87 | ||
| BC028526.1 | - | 1 | 73 | 3371464 | ||
| BY347589.1 | - | 2 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anapc13| Gene Symbol: Anapc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181394.3 | NP_852059.1 | 2 | 108 | ||
| GenBank mRNA | AK007332.1 | - | 1 | 87 | ||
| BC028526.1 | - | 1 | 73 | 3371464 | ||
| BY347589.1 | - | 2 | 108 | |||