siRNA Details
| Gene Symbol: Spcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026911.3 | NP_081187.2 | 2, 3 | 530 | ||
| GenBank mRNA | AI848281.1 | - | 2, 3 | 489 | ||
| AK007336.1 | - | 2, 3 | 228 | |||
| AK082928.1 | - | 2, 3 | 229 | |||
| BC034270.2 | - | 2, 3 | 405 | |||
| BC049895.1 | - | 2, 3 | 221 | 3991764 | ||
| BX512708.1 | - | 2, 3 | 472 | |||

Spcs1
Genomic Map