siRNA Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026917.5 | NP_081193.2 | 4 | 749 | ||
| GenBank mRNA | AB088044.1 | - | 3 | 515 | ||
| AK007355.1 | - | 5 | 1221 | |||
| AK018755.1 | - | 4 | 715 | |||
| AK080541.1 | - | 4 | 715 | |||
| AK169197.1 | - | 696 | ||||
| AY166672.1 | - | 3 | 515 | |||
| BC005531.1 | - | 5 | 776 | |||
| BC018334.1 | - | 3 | 585 | |||
| BC025126.1 | - | 4 | 734 | 3669723 | ||
| BC052464.1 | - | 4 | 654 | 6834763 | ||

Zdhhc3
Genomic Map