siRNA Details
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026917.5 | NP_081193.2 | 6 | 950 | ||
GenBank mRNA | AB088044.1 | - | 5 | 716 | ||
AK007355.1 | - | 7 | 1422 | |||
AK080541.1 | - | 6 | 916 | |||
AK169197.1 | - | 897 | ||||
AY166672.1 | - | 5 | 716 | |||
BC005531.1 | - | 7 | 977 | |||
BC018334.1 | - | 5 | 786 | |||
BC025126.1 | - | 6 | 935 | 3669723 | ||
BC052464.1 | - | 6 | 855 | 6834763 |
