siRNA Details
| Gene Symbol: Chac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026929.4 | NP_081205.1 | 3 | 680 | ||
| GenBank mRNA | AK007378.1 | - | 3 | 633 | ||
| AK032218.1 | - | 3 | 666 | |||
| BC025169.1 | - | 3 | 621 | 4483043 | ||
| BI872476.1 | - | 3 | 557 | |||

Chac1
Genomic Map