siRNA Details
| Gene Symbol: 1810010H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163473.1 | NP_001156945.1 | 2 | 223 | ||
| GenBank mRNA | BC124997.1 | - | 1 | 177 | ||
| BC125006.1 | - | 1 | 177 | |||
| BY099414.1 | - | 2 | 223 | |||
| CJ236692.1 | - | 2 | 193 | |||

1810010H24Rik
Genomic Map