siRNA Details
Gene Symbol: 1810011O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026931.2 | NP_081207.1 | 1 | 193 | ||
GenBank mRNA | AK007441.1 | - | 1 | 194 | ||
BC016562.1 | - | 1 | 150 | 3154083 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044104.1 | - | 398 | Chr. - - [-] on Build |
