siRNA Details
| Gene Symbol: 1810011O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026931.2 | NP_081207.1 | 1 | 238 | ||
| GenBank mRNA | AK007441.1 | - | 1 | 239 | ||
| BC016562.1 | - | 1 | 195 | 3154083 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK044104.1 | - | 353 | Chr. - - [-] on Build | ||

1810011O10Rik
Genomic Map