siRNA Details
| Gene Symbol: Gmppa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133708.3 | NP_598469.1 | 11 | 1045 | ||
| GenBank mRNA | AK007463.1 | - | 11 | 1030 | ||
| AK144311.1 | - | 11 | 1310 | |||
| AK150473.1 | - | 12 | 1181 | |||
| BC008116.1 | - | 11 | 1004 | 3498021 | ||

Gmppa
Genomic Map