siRNA Details
| Gene Symbol: Gmppa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133708.3 | NP_598469.1 | 5 | 497 | ||
| GenBank mRNA | AK007463.1 | - | 5 | 488 | ||
| AK144311.1 | - | 5 | 762 | |||
| AK150473.1 | - | 6 | 633 | |||
| AK184824.1 | - | 1 | 60 | |||
| AK186320.1 | - | 1 | 60 | |||
| AK189243.1 | - | 1 | 60 | |||
| AK190069.1 | - | 1 | 60 | |||
| BC008116.1 | - | 5 | 456 | 3498021 | ||

Gmppa
Genomic Map