siRNA Details
| Gene Symbol: Vps26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178027.4 | NP_821170.1 | 3 | 887 | ||
| GenBank mRNA | AK010160.1 | - | 3 | 821 | ||
| AK031589.1 | - | 3 | 890 | |||
| AK035321.1 | - | 3 | 840 | |||
| AK150929.1 | - | 3 | 808 | |||
| AK155578.1 | - | 3 | 848 | |||
| AK165539.1 | - | 3 | 826 | |||
| AK171154.1 | - | 3 | 846 | |||
| BC049180.1 | - | 3 | 850 | 6314347 | ||
| BC052721.1 | - | 3 | 811 | 6809853 | ||
| BC062972.1 | - | 3 | 811 | 6809878 | ||

Vps26b
Genomic Map