siRNA Details
| Gene Symbol: Trim15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024134.2 | NP_001019305.1 | 5 | 900 | ||
| NM_001177872.1 | NP_001171343.1 | 5 | 923 | |||
| GenBank mRNA | AK007445.1 | - | 5 | 924 | ||
| BC027186.1 | - | 5 | 900 | 4215801 | ||

Trim15
Genomic Map