siRNA Details
| Gene Symbol: 1810009N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026939.1 | NP_081215.1 | 1503 | |||
| GenBank mRNA | AK007409.1 | - | 1503 | |||
| AK084779.1 | - | 989 | ||||
| BC051588.1 | - | 175 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810009N02Rik| Gene Symbol: 1810009N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026939.1 | NP_081215.1 | 1503 | |||
| GenBank mRNA | AK007409.1 | - | 1503 | |||
| AK084779.1 | - | 989 | ||||
| BC051588.1 | - | 175 | ||||