siRNA Details
| Gene Symbol: Cnot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026949.3 | NP_081225.1 | 4 | 558 | ||
| GenBank mRNA | AK007581.1 | - | 4 | 622 | ||
| AK075727.1 | - | 4 | 528 | |||
| AK150757.1 | - | 4 | 639 | |||
| AK153907.1 | - | 4 | 561 | |||
| AK216161.1 | - | 2 | 170 | |||
| AY515608.1 | - | 3 | 525 | |||
| BC004040.1 | - | 4 | 558 | 3496188 | ||

Cnot8
Genomic Map