siRNA Details
| Gene Symbol: Fam25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183278.2 | NP_899101.1 | 2, 3 | 165 | ||
| GenBank mRNA | AK008614.1 | - | 2, 3 | 165 | ||
| BC086894.1 | - | 2, 3 | 167 | |||
| BC147104.1 | - | 2, 3 | 154 | 9056159 | ||
| BC147105.1 | - | 2, 3 | 154 | 9056136 | ||

Fam25c
Genomic Map