siRNA Details
| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026956.2 | NP_081232.2 | 8 | 695 | ||
| GenBank mRNA | BC021323.1 | - | 8 | 776 | ||
| BC038121.1 | - | 8 | 1098 | |||
| BC046292.1 | - | 6 | 736 | |||
| BC052782.1 | - | 8 | 686 | |||
| BC099497.1 | - | 8 | 690 | |||

Cd209f
Genomic Map