siRNA Details
| Gene Symbol: Snx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080557.2 | NP_542124.1 | 10 | 917 | ||
| GenBank mRNA | AK007706.1 | - | 10 | 959 | ||
| AK031368.1 | - | 929 | ||||
| AK135356.1 | - | 10 | 924 | |||
| BC006043.1 | - | 10 | 907 | |||
| BC016599.1 | - | 10 | 917 | 2644991 | ||

Snx4
Genomic Map