siRNA Details
| Gene Symbol: Bola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026975.2 | NP_081251.1 | 2 | 189 | ||
| GenBank mRNA | AK007718.1 | - | 2 | 190 | ||
| BC027558.1 | - | 2 | 185 | 1477943 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bola1| Gene Symbol: Bola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026975.2 | NP_081251.1 | 2 | 189 | ||
| GenBank mRNA | AK007718.1 | - | 2 | 190 | ||
| BC027558.1 | - | 2 | 185 | 1477943 | ||