siRNA Details
| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028839.4 | NP_083115.3 | 1, 2 | 287 | ||
| GenBank mRNA | AK007740.1 | - | 1, 2 | 295 | ||
| AK009341.1 | - | 1, 2 | 266 | |||
| AK076341.1 | - | 1, 2 | 266 | |||
| AK149088.1 | - | 1, 2 | 260 | |||
| AK155257.1 | - | 1, 2 | 252 | |||
| CJ180452.1 | - | 1, 2 | 291 | |||

Tmem110
Genomic Map