siRNA Details
| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133710.3 | NP_598471.3 | 8 | 1051 | ||
| GenBank mRNA | AJ344340.1 | - | 8 | 1133 | ||
| AK013115.1 | - | 8 | 1047 | |||
| AK142857.1 | - | 3 | 834 | |||
| BC094289.1 | - | 8 | 736 | 4986338 | ||

Ctdspl
Genomic Map