siRNA Details
| Gene Symbol: Glipr1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027018.1 | NP_081294.1 | 2 | 417 | ||
| GenBank mRNA | AK005860.1 | - | 2 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glipr1l1| Gene Symbol: Glipr1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027018.1 | NP_081294.1 | 2 | 417 | ||
| GenBank mRNA | AK005860.1 | - | 2 | 418 | ||