siRNA Details
Gene Symbol: Glipr1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027018.1 | NP_081294.1 | 4 | 699 | ||
GenBank mRNA | AK005860.1 | - | 4 | 700 | ||
BX635028.1 | - | 2 | 197 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Glipr1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027018.1 | NP_081294.1 | 4 | 699 | ||
GenBank mRNA | AK005860.1 | - | 4 | 700 | ||
BX635028.1 | - | 2 | 197 |