siRNA Details
| Gene Symbol: Izumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027034.1 | NP_081310.1 | 1 | 255 | ||
| GenBank mRNA | BC099579.1 | - | 1 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Izumo3| Gene Symbol: Izumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027034.1 | NP_081310.1 | 1 | 255 | ||
| GenBank mRNA | BC099579.1 | - | 1 | 255 | ||