siRNA Details
| Gene Symbol: 1700001L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027035.1 | NP_081311.1 | 3 | 445 | ||
| GenBank mRNA | AK005609.1 | - | 3 | 446 | ||
| AK138387.1 | - | 3 | 446 | |||
| BC049693.1 | - | 4 | 355 | |||
| BC119515.1 | - | 3 | 432 | 40055598 | ||
| BC119516.1 | - | 3 | 431 | 40055603 | ||

1700001L19Rik
Genomic Map