siRNA Details
| Gene Symbol: 1700012B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162428.1 | NP_001155900.1 | 3 | 452 | ||
| NM_027038.1 | NP_081314.1 | 3 | 452 | |||
| GenBank mRNA | AK005888.1 | - | 3 | 446 | ||
| AK132924.1 | - | 453 | ||||
| AK160153.1 | - | 3 | 453 | |||
| BC048675.1 | - | 3 | 428 | 6774204 | ||
| BC099559.1 | - | 3 | 431 | |||

1700012B07Rik
Genomic Map