siRNA Details
| Gene Symbol: 1700007K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027040.1 | NP_081316.1 | 3 | 431 | ||
| GenBank mRNA | AK005731.1 | - | 3 | 441 | ||
| AY927869.1 | - | 3 | 376 | |||
| BC048581.1 | - | 3 | 412 | |||
| BC099566.1 | - | 3 | 431 | 30916995 | ||

1700007K13Rik
Genomic Map