siRNA Details
Gene Symbol: 1700008P02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027048.1 | NP_081324.1 | 2, 3 | 510 | ||
GenBank mRNA | AK005786.1 | - | 2, 3 | 512 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700008P02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027048.1 | NP_081324.1 | 2, 3 | 510 | ||
GenBank mRNA | AK005786.1 | - | 2, 3 | 512 |