siRNA Details
| Gene Symbol: 1700008O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027049.1 | NP_081325.1 | 4 | 351 | ||
| GenBank mRNA | AK005784.1 | - | 4 | 352 | ||
| BC127243.1 | - | 2 | 467 | |||
| BC127244.1 | - | 2 | 467 | |||
| BF467486.1 | - | 3 | 572 | |||

1700008O03Rik
Genomic Map