siRNA Details
| Gene Symbol: Smim23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027050.1 | NP_081326.1 | 1 | 124 | ||
| GenBank mRNA | AK005748.1 | - | 1 | 104 | ||
| EU477371.1 | - | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim23| Gene Symbol: Smim23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027050.1 | NP_081326.1 | 1 | 124 | ||
| GenBank mRNA | AK005748.1 | - | 1 | 104 | ||
| EU477371.1 | - | 1 | 47 | |||